Calibration weights, OIML M1
10 g, With certificate
Catalog # OHAU11117223
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
Delivery information: Supplied in a plastic box.
Documents
OHAU11117232_TDS_EN
Datablad
Specifications
- Weight:10 g
- Seq:04
- Type:With certificate