Kalibreringslodder, OIML M1
50 g, Uden certifikat
Catalog # OHAU11117146
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
Delivery information: Supplied in a plastic box.
Documents
OHAU11117232_TDS_EN
Datasheet
Specifications
- Vægt:50 g
- Seq:19
- Model:Uden certifikat