Calibration weights, OIML M1
10 g, Without certificate
Catalog # OHAU11117144
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
: Supplied in a plastic box.
Specifications
- Weight:10 g
- Seq:17
- Type:Without certificate