D-(+)-Glucose monohydrate
Glass bottle for solids, 2.500 g
CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450
: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- :14431-43-7
- :C₆H₁₂O₆·H₂O
- :MFCD00149450
- :198.17 g/mol
- :∼ 83 °C
- :Ambient
- :200-075-1
- Pack type:Glass bottle for solids
- Pk:2.500 g