Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test tubes with screw caps

Test tubes with screw caps

13 ml, 100 mm, 16 mm, Natural, Round bottom, screw cap enclosed, non sterile
Catalog # 212-0508
Supplier:   BOETTGER PAUL
Test tubes with screw caps
Catalog # 212-0508
Supplier:  BOETTGER PAUL
Test tubes with screw caps
Test tubes with screw caps

13 ml, 100 mm, 16 mm, Natural, Round bottom, screw cap enclosed, non sterile

Price
1.220,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

PP, transparent.

  • Good chemical and temperature resistance
  • Tight sealing screw cap made from PE
  • Available with enclosed or fitted screw cap, standard-hygienic or sterile

For safe transportation and storage of samples.

Specifications
  • Ext.Ø:
    16 mm
  • Type:
    non sterile
  • Capacity:
    13 ml
  • Description:
    Round bottom, screw cap enclosed
  • Length:
    100 mm
  • Seq:
    05
  • Cap colour:
    Natural
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.