[EN]CONICAL INSERT 100UL 5X30 MM 1 * 100 ST
Catalog # BROW150610
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
[EN]CONICAL INSERT 100UL 5X30 MM 1 * 100 ST
Catalog # BROW150610
Supplier: BROWN CHROMATOGRAPHY SUPPLIES
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Syringe filters, Acrodisc® One, wwPTFE membrane
Supplier: Cytiva (Formerly Pall Lab)
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.