Order Entry
Denmark
Orders LinkContactUsLinkComponent
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Black modules, framed, C8, PolySorp surface, Black, 96
Catalog # 738-0751
   Low Manufacturing Impact
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Catalog # 738-0751
   Low Manufacturing Impact
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Modules, Immuno™ LockWell™ Fluoronunc™/Luminunc™
Black modules, framed, C8, PolySorp surface, Black, 96
Price
3.590,00 kr
Log in for availability
1 * 60 Items
Product Details & Documents
PS, 96 wells per frame. For fluorescence/luminescence-based immunoassays and binding assays, these modules have been designed using the standard 96 MicroWell™ LockWell™ format. Opaque white modules have a reflective surface and offer low cross talk and minimal background. Opaque black modules with light absorbing surface offer minimum cross talk and back scattered light.
  • Each well locks in the frame of the same height to prevent jamming in machines
  • Easy to handle
  • Uniform washing and reading
  • Certified binding homogeneity for all four surfaces
  • MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules
Total volume per well: 350 µl
Specifications
  • No. of wells:
    96
  • Colour:
    Black
  • Description:
    Black modules, framed, C8, PolySorp surface
  • Cat. No.:
    738-0751
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact