Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder pillows, iron
Iron (FerroMo™) reagent 2, 0,01 - 1,80 mg/l, FerroMo™, -, PP, 50
Katalog # HACH2543866
Leverandør:   Hach
Powder pillows, iron
Katalog # HACH2543866
Leverandør:  Hach
CAS nummer:  
Powder pillows, iron
Powder pillows, iron
Iron (FerroMo™) reagent 2, 0,01 - 1,80 mg/l, FerroMo™, -, PP, 50
CAS nummer:  
Price
411,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Specifikationer
Specifikationer
  • Chemical method:
    FerroMo™
  • Pk:
    50
  • Description:
    Iron (FerroMo™) reagent 2
  • Sample size:
    -
  • Type:
    PP
  • Range:
    0,01 - 1,80 mg/l
Specifikationer