Plasmid DNA buffers
DNA wash buffer concentrate, low salt spin column was solution (makes 500 ml), 100 mL
Supplier: OMEGA BIO-TEK
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pk:100 mL
- Description:DNA wash buffer concentrate, low salt spin column was solution (makes 500 ml)



