Programmable Vortex Mixers, Genie Series
Product Details & Documents
Same great features of the Original Vortex-Genie® 2 in addition to programmability. These rugged and reliable vortex mixers have a durable motor, solid metal casing and rubber feet that prevent them 'walking'. The Vortex-Genie® Pulse is ideal for applications requiring more aggressive action than standard vortexing.
- These vortex mixers provide accuracy, reproducibility and repeatability
- Models have digital control and display of speed, time, and 'RUN/STOP' time, for reproducibility and accuracy, with true speed display
- Can also be used as standard 'continuous' vortex mixers
- Vortex-Genie® Pulse’s true pulsing action (interval mixing) gives more collisions and random motion
- Variable 'RUN/STOP' time provides quick pulsing or interval mixing with Vortex-Genie® Pulse
- 2 year warranty
Digital true vortex mixing as incorporated in the original Vortex-Genie. Programmability achieves accurate and repeatable results, especially for use with kits or repetitive protocols. The pulse feature of the Vortex-Genie Pulse allows rapid pulsing or timed “on/off” mixing for interval mixing, with the benefits of interruption of the vortex to achieve better mixing.
Both units accept all Vortex-Genie 2 accessories. Optimized for use with high speed attachments/accessories.
Delivery information: The Digital Vortex-Genie 2 and Vortex-Genie Pulse are supplied with the standard pop-off cup for single test tubes and a standard 76 mm platform suitable for beakers, flasks and multiple test tubes. The ABI Digital Vortex-Genie 2 is supplied with microplate adaptor and bands. Vortex-Genie® Pulse is supplied with quick change standard pop-off cup for single test tubes and 76 mm platform suitable for beakers, flasks and multiple tests tubes. Unit can be used with other optional Vortex-Genie accessories, which must be purchased separately
Caution: * Depending on speed
** Multiple sample starter set consists of a 152 mm platform, a foam insert for 60 microtubes and a foam insert for 1 standard microplate
*** Large sample set is ideal for mixing beakers, flasks and large tubes. The set consists of a recessed platform, 2 elastic bands, a blank foam insert, a foam insert for 4× 29 − 37 mm tubes, a foam insert for 8× 14 − 29 mm tubes and a foam insert for 12× 9 − 16 mm tubes
**** Small ampoule/tube attachment enables vigorous end-to-end mixing for up to four 15 − 17 mm Ø tubes or ampoules
***** Large ampoule/tube attachment enables vigorous end-to-end mixing for up to four 10 − 17 mm Ø tubes or ampoules
Specifikationer
- Description:Digital Vortex-Genie® 2, for Applied Biosystems microplate protocols, EU plug
- Seq:011
- Shaking motion:Orbital
- Cat. No.:444-0874
- Max. capacity:500 ml
- Orbit:3,5 mm
- Timer:1 - 99 min or continuous
- Speed range:500 - 2850 min⁻¹
- Model:Digital Vortex-Genie® 2
- W×D×H:122×165×165 mm
- Power supply:230 V
- Weight:4 kg
Accessories
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1049Vertical tube holder, 50 ml (for 6×50 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 1.810,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5919Support for microtitre plates (fits in 152 mm platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 786,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCII504-0232-00Deepwell microplate foam insert (2). Require 6-inch platformVortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 2 Items - 962,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5905Platform, 152 mm, without insert (inserts listed below)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 643,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5904Multiple sample starter set*Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 1.430,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5916Recessed platform (foam inserts listed below)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 1.740,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1052Vertical microtube holder, 15 ml and 50 ml (for 6×50 ml, 6×15 ml tubes and 12 microtubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.500,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1047Vertical high speed tube holder, 15 ml (for 6×15 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 1.910,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5902Platform, Ø 75 mm without rubber coverVortex-Genies®Log ind for tilgængelighed1 * 1 Items - 456,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5914Foam adapter for tubes 9 − 16 mm Ø (fits in recessed platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 358,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-0775Holder for 12×2,0 ml screw-cap microtubes (fits on TurboMix attachment)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 1.030,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1046Vertical high speed tube holder, 50 ml (for 3×50 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 1.680,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-9162Large ampoule/tube attachment****Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 2.540,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1043Horizontal tube holder, 50 ml (for 6×50 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 1.810,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCII0K-0513-900Replacement elastic bandsVortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Each - 107,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5917Assorted tube inserts: 1 each of the following: blank, 9 − 16 mm, 14 − 29 mm and 29 − 37 mm (fit in recessed platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 4 Items - 732,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCIISI-0513Microplate adapter for applied biosystemsVortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 1.740,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5903Rubber cover for Ø 75 mm platformVortex-Genies®Log ind for tilgængelighed1 * 1 Items - 448,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-9161Small ampoule/tube attachment***Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 1.740,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1051Vertical microtube holder (for 24 microtubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.290,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1048Vertical high speed microtube holder (for 9 microtubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.130,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5915Elastic bands to help secure vessels onto recessed platformVortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 107,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5912Foam adapter for tubes 14 − 29 mm Ø (fits in recessed platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 358,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5913Foam insert, blank (without holes) (fits in recessed platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 358,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5911Foam adapter for tubes 29 − 37 mm Ø (fits in recessed platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 358,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-0061Two-tier insert for 96 well plate (fits in 152 mm platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 1 Items - 891,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCII0K-0236-408Bottom closure with screw kitVortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 640,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- SCIISI-HP524Horizontal plastic clip microtube holder (for 24 microtubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.240,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1050Vertical tube holder, 15 ml (for 12×15 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.060,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5901Pop-off cupVortex-Genies®, Disruptor-GenieLog ind for tilgængelighed1 * 1 Items - 448,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1045Horizontal microtube holder (for 24 microtubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.290,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-1044Horizontal tube holder, 15 ml (for 12×15 ml tubes)Vortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 2.060,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-0724TurboMix attachment for 12×1,5/2,0 ml snap-top or screw-cap microtubesVortex Genie® 2, Vortex PulseLog ind for tilgængelighed1 * 1 Items - 3.520,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-5921Adapter for 60 microtubes (fits in 152 mm platform)Vortex-Genies® except Vortex-Genie® 1Log ind for tilgængelighed1 * 2 Items - 768,00 kr
Produktinformation
- Katalog #DescriptionForTilgængelighedPrice
- 444-0443Platform with rubber cover, 3 inchVortex-Genie® 2, Vortex-Genie® pulseLog ind for tilgængelighed1 * 1 Items - 713,00 kr