Span® 80, Supelco®
Glass bottle, 250 mL
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Arlacel 80, Sorbitan monooleate
- CAS Number:1338-43-8
- Formula:C₂₄H₄₄O₆
- MDL Number:MFCD00080948
- Molecular Weight:428.61 g/mol
- Density:1 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:215-665-4
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



