Triethylammonium acetate buffer, BioUltra, volatile buffer, Fluka®
Triethylammonium acetate buffer, 1,0 M in H₂O, 7,0, ±0,1, Glass bottle, 500 mL
Supplier: Honeywell Chemicals
Triethylammonium acetate buffer, BioUltra, volatile buffer, Fluka®
Catalog # 90357-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
- Absorbance 260 nm: max. 0,03
- Absorbance 280 nm: max. 0,03
Documents
Specifications
Specifications
- Concentration:1,0 M in H₂O
- Pack type:Glass bottle
- Pk:500 mL
- Description:Triethylammonium acetate buffer
- pH accuracy:±0,1
- pH value:7,0
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



