Triethylammonium acetate buffer, BioUltra, volatile buffer, Fluka®
Triethylammonium acetate buffer, 1,0 M in H₂O, 7,0, ±0,1, Glass bottle, 100 mL
Triethylammonium acetate buffer, BioUltra, volatile buffer, Fluka®
90357-100ML
:
- Absorbance 260 nm: max. 0,03
- Absorbance 280 nm: max. 0,03
Documents
- Concentration:1,0 M in H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Description:Triethylammonium acetate buffer
- pH accuracy:±0,1
- pH value:7,0
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



