Supelco® Organic Reference Standard, Dextran
Dextran, Analytical standard, for GPC, 5000, 500 mg
Katalog # 00269-500MG
Leverandør: Merck
Jump to:
Product Details & Documents
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Specifications
Specifications
- Pk:500 mg
- Standards:Analytical standard, for GPC, 5000
- Description:Dextran
- CAS No.:9004-54-0
- Formula:[C₆H₁₀O₅]ₙ
- Flash point:Not applicable
- Shipping conditions:STD
- MDL No.:MFCD00130935
- Cat. No.:00269-500MG
- M-Clarity quality level:MQ100
- Storage conditions:2...30 °C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



