Supelco® Organic Reference Standard, Dextran
Dextran, Analytical standard, for GPC, 5000, 100 mg
00269-100MG
: Merck
: Bottomless glass bottle. Contents are inside inserted fused cone.
- Pk:100 mg
- Standards:Analytical standard, for GPC, 5000
- Description:Dextran
- Cat. No.:00269-100MG
- M-Clarity quality level:MQ100
- Formula:[C₆H₁₀O₅]ₙ
- Storage conditions:2...30 °C
- MDL No.:MFCD00130935
- Flash point:Not applicable
- CAS No.:9004-54-0
- Shipping conditions:STD



