for NON Vacuum autoclaves 1 * 10 items
Catalog # TUTTCLE096-0026
Supplier: TUTTNAUER
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Chamber Brite pack tablets Autoclave Cleaner 1 * 12 items
Supplier: TUTTNAUER
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals