Iron ore
NIST-693 Iron ore, nimba - constituents, Iron, Fe, 100 g
Catalog # PROONIST-693
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
This reference standard is intended for use in checking chemical methods of analysis and in the calibration with instrumental methods of analysis.
Certifications: Traceable to NIST.
Specifications
Specifications
- Pk:100 g
- Description:NIST-693 Iron ore, nimba - constituents
- Element:Iron
- Symbol:Fe