Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide, micropearls for analysis

Sodium hydroxide, micropearls for analysis

Plastic bottle, 1 kg
Catalog # ACRO383040010
Corrosive
Sodium hydroxide, micropearls for analysis
Catalog # ACRO383040010
CAS Number:  1310-73-2
Sodium hydroxide, micropearls for analysis
Sodium hydroxide, micropearls for analysis

Plastic bottle, 1 kg

CAS Number:  1310-73-2
Price
529,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:318 °C
  • Density:2,13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27

Documents

Specifications

Specification Test Results

  • Appearance:
    White Pearls
  • Titration with HCl:
    ≥99.0 %
  • Sodium carbonate (Na2CO3):
    ≤0.4 %
  • Sodium sulfate (Na2SO4):
    ≤80 ppm
  • Sodium chloride (NaCl):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Lead (Pb):
    ≤0.5 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications