Sodium L(+)-ascorbate 99%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Sodium L(+)-ascorbate 99%
Catalog # ACRO352681000
Supplier: Thermo Fisher Scientific
CAS Number: 134-03-2
Jump to:
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219…221 °C (dec.)
- Density:1,799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Documents
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Iodimetric:98.5 to 101.5 %
- Solubility:(5% in water) Clear colorless to light yellow
- Specific optical rotation:+103° to +108° (20°C, 589 nm) (c=5, H2O)
Specifications
- Pack type:Plastic bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



