Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium L(+)-ascorbate 99%

Sodium L(+)-ascorbate 99%

Glass bottle for solids, 5 g
Catalog # ACRO352680050
Sodium L(+)-ascorbate 99%
Catalog # ACRO352680050
CAS Number:  134-03-2
Sodium L(+)-ascorbate 99%
Sodium L(+)-ascorbate 99%

Glass bottle for solids, 5 g

CAS Number:  134-03-2
Price
258,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Melting Pt:219…221 °C (dec.)
  • Density:1,799 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-126-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Iodimetric:
    98.5 to 101.5 %
  • Solubility:
    (5% in water) Clear colorless to light yellow
  • Specific optical rotation:
    +103° to +108° (20°C, 589 nm) (c=5, H2O)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications