Resorcinol 98%
Plastic bottle, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol
- CAS Number:108-46-3
- Formula:C₆H₆O₂
- MDL Number:MFCD00002269
- Molecular Weight:110.11 g/mol
- Merck Index:12,08323
- Boiling Pt:281 °C (760 mmHg)
- Melting Pt:109…111 °C
- Density:1,272 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Ambient
- EINECS:203-585-2
- UN:2876
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to yellow to beige to pink Crystals or granules or flakes
- Infrared spectrum:Conforms
- Melting point:108 to 112 °C
- GC:≥97.5 %
Specifications
- Pack type:Plastic bottle
- Pk:50 g



