Cyclohexane 99+%, pure
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Cyclohexane 99+%, pure
Catalog # ACRO111110100
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:6.5 °C
- Density:0.77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
ACRO11111
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (K.F.)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
Specifications
- Pack type:Plastic drum
- Pk:10 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



