Order Entry
Denmark
Orders LinkContactUsLinkComponent
Mouse single tissue blot, single species
Mouse single tissue blot, single species
Mouse normal lung single tissue blot, 1
Katalog # TB05
Leverandør:   G-Biosciences
Mouse single tissue blot, single species
Katalog # TB05
Leverandør:  G-Biosciences
CAS nummer:  
Mouse single tissue blot, single species
Mouse single tissue blot, single species
Mouse normal lung single tissue blot, 1
CAS nummer:  
Price
3.140,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue mouse blots
Specifikationer
Specifikationer
  • Pk:
    1
  • Description:
    Mouse normal lung single tissue blot
Specifikationer