G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH), 1 gallon
Catalog # R016
Supplier: G-Biosciences
G-Biosciences Hybridization Buffer (Hybridization Denaturing Solution)
Catalog # R016
Supplier: G-Biosciences
CAS Number:
Jump to:
Product Details & Documents
Ready-to-Use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
1.5 M sodium chloride
- 0.5 M sodium hydroxide
A high-pH reagent used to denature double-stranded nucleic acids into single strands, preparing DNA or RNA for hybridization. Ready-to-use: Convenient, time-saving formulation ideal for consistent experimental reproducibility.
Specifications
Specifications
- Pk:1 gallon
- Description:Hybridization denaturing solution (1,5 M NaCl, 0,5 N NaOH)