RNaseOUT™ RNAse removal solution
RNaseOut™, refill, 1 L
786-71
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
- Simply spray and rinse
- RNaseOUT™ is non-toxic and residue free
- RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work
- Pk:1 L
- Description:RNaseOut™, refill