Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffer solutions, according to DIN 19266, Fluka™

Buffer solutions, according to DIN 19266, Fluka™

9,180, —, Borate, 25 °C, Plastic bottle, 1×1 L, 1 L
Catalog # 33595-1L
Supplier:   Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Catalog # 33595-1L
CAS Number:  
Buffer solutions, according to DIN 19266, Fluka™

9,180, —, Borate, 25 °C, Plastic bottle, 1×1 L, 1 L

CAS Number:  
Price
517,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Buffers with fungicide (except pH 10,012)

Certifications: According to DIN 19266.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • pH accuracy:
  • Pk Info:
    1×1 L
  • pH value:
    9,180

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.