Dropping funnels, cylindrical, without pressure balance
50 ml, 19/26 NS, 19/26 NS, 1 ml
Katalog # WITG1.802.050
Leverandør: witeg Labortechnik
Dropping funnels, cylindrical, without pressure balance
Katalog # WITG1.802.050
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Cylindrical dropping funnels, made of borosilicate glass with screw-thread retaining nut.
- With glass key stopcock and graduation
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Certifications: DIN 12567
Specifikationer
- Cone size:19/26 NS
- Socket size:19/26 NS
- Capacity:50 ml
- Division:1 ml
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



