Order Entry
Denmark
Orders LinkContactUsLinkComponent
FlashGel™ RNA system

FlashGel™ RNA system

FlashGel™ RNA cassettes, 1,2% agarose, 12+1 single tier
Catalog # 733-1617
Supplier:   LONZA
FlashGel™ RNA system
Catalog # 733-1617
Supplier:  LONZA
FlashGel™ RNA system
FlashGel™ RNA system

FlashGel™ RNA cassettes, 1,2% agarose, 12+1 single tier

Price
2.760,00 kr
Log in for availability
1 * 9 Items
Product Details & Documents

FlashGel™ system for RNA is optimised for the unique requirements of RNA, and is an ideal tool for rapid analysis of sample integrity. High quality, intact RNA is essential for consistent results in gene expression, Northern analysis, cDNA library construction and cDNA labelling for microarrays. Most protocols recommend checking RNA prior to downstream analysis.

  • Complete RNA analysis in less than 30 minutes
  • Detects <10 ng RNA per band, separation range 0,5 to 9 Kb
  • Avoids the use of hazardous reagents and contaminating RNases

Certifications: Some components and technology of the FlashGel™ system are sold under licensing agreements. The nucleic acid stain in this product is manufactured and sold under license from Molecular Probes, Inc for use only in research applications or quality control, and is covered by pending and issued patents. The FlashGel Dock technology is licensed from Clare Chemical Research, Inc. The electrophoresis technology is licensed from Temple University.

Delivery information: Supplied as separate components or as a starter kit. Starter kit contains box of 9, 1,2%, 12+1 well FlashGel™ RNA cassettes; sample buffer; RNA marker; and molecular biology grade water. FlashGel™ dock sold separately.

Caution: Gel cannot be accessed. Unsuitable for recovery.

Specifications
  • Description:
    FlashGel™ RNA cassettes, 1,2% agarose, 12+1 single tier
  • Seq:
    09
  • Cat. No.:
    733-1617
  • Separation range (bp):
    0,5 kb - 9 kb
  • Cassette size W×D×H:
    107×115×17 mm
  • Dock size W×D×H:
    120×134×54 mm
  • Gel size W×L:
    84×70 mm
  • Storage (cassettes):
    Room temperature for 3 months from date of manufacture
  • Model:
    FlashGel™ RNA
  • Well volume:
    2 - 5 µl
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ power supply
    FlashGel™ power supply
    LONZ57068
    Log in for availability
    1 * 1 Each - 10.050,00 kr

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ power supply pack, includes FlashGel™ dock
    FlashGel™ power supply pack, includes FlashGel™ dock
    LONZ57069
    Log in for availability
    1 * 1 Each - 19.780,00 kr

Product Information

  • Catalog #
    Availability
    Price
  • FlashGel™ dock
    FlashGel™ dock
    733-1609
    Log in for availability
    1 * 1 Items - 10.960,00 kr

Product Information

  • Catalog #
    Availability
    Price
  • RNA marker (0,5 bp to 9 Kb)
    RNA marker (0,5 bp to 9 Kb)
    733-1734
    Log in for availability
    1 * 1 Each - 6.390,00 kr

Product Information

  • Catalog #
    Availability
    Price
  • Formaldehyde sample buffer
    Formaldehyde sample buffer
    733-1739
    Log in for availability
    1 * 5 mL - 969,00 kr

Product Information

  • Catalog #
    Availability
    Price
  • RNA marker (0,5 - 9 kb), 50 µL
    RNA marker (0,5 - 9 kb), 50 µL
    733-1623
    Log in for availability
    1 * 50 µl - 3.770,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.