Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spouts for jerricans
Spouts for jerricans
Flexible spout with safety vent, S70/71
Katalog # 590-0849
Leverandør:   S.C.A.T.
Spouts for jerricans
Katalog # 590-0849
Leverandør:  S.C.A.T.
Spouts for jerricans
Spouts for jerricans
Flexible spout with safety vent, S70/71
Price
132,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
For simple and safe pouring of fluids from containers.
  • Plastic
  • Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Specifikationer
  • Thread:
    S70/71
  • Description:
    Flexible spout with safety vent
  • Seq:
    06