Spouts for jerricans
Flexible spout with safety vent, S55
Catalog # 590-0845
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
For simple and safe pouring of fluids from containers.
- Plastic
- Suitable for S.C.A.T. safety containers
Safety Spouts are also suitable for politainer waste containers and are available in flexible and rigid versions. Designed for the common canister threads S55, S60/61, and S40/71, and made of high-quality PEHD or electrically conductive PEHD-EC material with additional ventilation.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Thread:S55
- Description:Flexible spout with safety vent
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



