Order Entry
Denmark
Orders LinkContactUsLinkComponent
Saccharometers

Saccharometers

40 - 50, 0,1, 260 mm
Catalog # AMARH845036
Supplier:   Amarell
Saccharometers
Catalog # AMARH845036
Supplier:  Amarell
Saccharometers
Saccharometers

40 - 50, 0,1, 260 mm

Price
272,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Saccharometers with reference temperature of 20 °C.

Specifications
  • Length:
    260 mm
  • Seq:
    05
  • Division:
    0,1
  • Range:
    40 - 50
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.