Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
45/40, 65 mm
  COWI010.1245
 :   COWIE
Stoppers, self realasing, PTFE
  COWI010.1245
 :  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
45/40, 65 mm
635,00 kr
1 * 1 Items
Fit standard ground glass joints.
With sealing rings and self releasing ring.
  • NS:
    45/40
  • Height:
    65 mm