Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
24/29, 55 mm
Catalog # COWI010.1224
Supplier:   COWIE
Stoppers, self realasing, PTFE
Catalog # COWI010.1224
Supplier:  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
24/29, 55 mm
Price
285,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifications
  • NS:
    24/29
  • Height:
    55 mm