Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
19/26, 50 mm
Catalog # COWI010.1219
Supplier:   COWIE
Stoppers, self realasing, PTFE
Catalog # COWI010.1219
Supplier:  COWIE
Stoppers, self realasing, PTFE
Stoppers, self realasing, PTFE
19/26, 50 mm
Price
215,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifications
  • NS:
    19/26
  • Height:
    50 mm