Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety Bottle Container

Safety Bottle Container

Silicone seal for transport container, Red, Container with Ø 160 mm
Catalog # BOCH18301SS
Supplier:   BOCHEM
Safety Bottle Container
Catalog # BOCH18301SS
Supplier:  BOCHEM
Safety Bottle Container
Safety Bottle Container

Silicone seal for transport container, Red, Container with Ø 160 mm

Price
kr234.00
Log in for availability
1 * 5 Items
Product Details & Documents

Safety transport container, made of stainless steel, for laboratory bottles.

  • For bottles 50 to 1000 ml
  • With hinge, lock and silicone seal
  • Good chemical resistance
Specifications
  • For:
    Container with Ø 160 mm
  • Colour:
    Red
  • Description:
    Silicone seal for transport container
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.