Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class 1
VWR®, Calibration Weights, Class 1
10000 g, 25000 ± mg
Catalog # 611-8766
Supplier:   VWR Collection
VWR®, Calibration Weights, Class 1
Catalog # 611-8766
Supplier:  VWR Collection
VWR®, Calibration Weights, Class 1
VWR®, Calibration Weights, Class 1
10000 g, 25000 ± mg
Price
10.300,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
Specifications
  • Weight:
    10000 g
  • Seq:
    08
  • Tolerance:
    25000 ± mg