Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR®, Self-wringing Mopping System
VWR®, Self-wringing Mopping System
Blue polyester sponge (406×127 mm) for use in corners with disinfectants
Catalog # 129-2502
Supplier:   VWR Collection
VWR®, Self-wringing Mopping System
Catalog # 129-2502
Supplier:  VWR Collection
VWR®, Self-wringing Mopping System
Blue polyester sponge (406×127 mm) for use in corners with disinfectants
Price
6.620,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
This mopping system can be used without a wringer. It is ideal for applying disinfectants to floors, walls and ceilings. The high performance foam for cleanrooms enables excellent, lasting liquid transfer.
  • Autoclavable; resistant to gamma rays and ETO
  • Resistant to the most widely available disinfectants and detergents
  • Sponges made from laminated polyester, for liquid absorption and highly delicate disinfection
  • Replacement foam mop heads are available with stainless steel or galvanised steel clips
Specifications
  • Description:
    Blue polyester sponge (406×127 mm) for use in corners with disinfectants
  • Seq:
    16