Order Entry
Denmark
ContactUsLinkComponent
Stirrer 20 x10 mm with permanent magnetic core and thick, white ptfe coating. with smooth surfaces and rounded ends. chemical and te mperature resistant up to +275 c. 1 * 1 items
Catalog # SEMA0266
Supplier:  SEMADENI
Stirrer 20 x10 mm with permanent magnetic core and thick, white ptfe coating. with smooth surfaces and rounded ends. chemical and te mperature resistant up to +275 c. 1 * 1 items
Catalog # SEMA0266
Supplier:  SEMADENI
Frequently Bought Together