Order Entry
Denmark
Orders LinkContactUsLinkComponent
Accessories for DOSE IT

Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 1 mm
Catalog # 710-1023
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1023
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 1 mm

Price
281,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Documents

Specifications
  • Description:
    Silicone tubing, autoclavable, length: 2,5 m
  • Ø int.:
    1 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.