Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR®, Cork Plates

VWR®, Cork Plates

200×200 mm
Catalog # 217-1009
Supplier:   VWR Collection
VWR®, Cork Plates
Catalog # 217-1009
Supplier:  VWR Collection
VWR®, Cork Plates
VWR®, Cork Plates

200×200 mm

Price
446,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents

Made of natural cork.

  • Square form
  • Thickness 15 mm
Specifications
  • W×D:
    200×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.