Stirrer guides
Stirrer guide, GTS-vacuum-stirrer, with PTFE-insert, —, 10,01 mm
WITG1.468.004
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.
- NS:—
- Description:Stirrer guide, GTS-vacuum-stirrer, with PTFE-insert
- Seq:02
- Shaft Ø:10,01 mm



