Order Entry
Denmark
ContactUsLinkComponent
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and UK-plug, Chamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # 481-2063
Supplier:  TUTTNAUER
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and UK-plug, Chamberdimension (xL) 380 x 690 mm Volume: 85 liters 1 * 1 items
Catalog # 481-2063
Supplier:  TUTTNAUER
Frequently Bought Together