Phthalic acid, bis-iso-octyl ester (technical) certified 1 * 0,25 g
Catalog # EHERC16173650
Supplier: EHRENSTORFER
Phthalic acid, bis-iso-octyl ester (technical) certified 1 * 0,25 g
Catalog # EHERC16173650
Supplier: EHRENSTORFER
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions
Supplier: REAGECON
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Diethyl phthalate
Supplier: EHRENSTORFER