Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 4 mm
Catalog # 710-1026
Supplier:   INTEGRA Biosciences
Accessories for DOSE IT
Catalog # 710-1026
Accessories for DOSE IT
Accessories for DOSE IT

Silicone tubing, autoclavable, length: 2,5 m, 4 mm

Price
329,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

 

Specifications
  • Description:
    Silicone tubing, autoclavable, length: 2,5 m
  • Ø int.:
    4 mm