Accessories for DOSE IT
Silicone tubing, autoclavable, length: 25 m (bulk roll), 3 mm
Catalog # 710-1031
Supplier: INTEGRA Biosciences
Jump to:
Product Details & Documents
Documents
Dose It
Videos
Specifications
- Description:Silicone tubing, autoclavable, length: 25 m (bulk roll)
- Ø int.:3 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



