Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Automatic burettes, Sofnol

10 ml, 0,1 ml, 0,01 ± ml
  612-2905
 :   SciLabware
Automatic burettes, Sofnol
  612-2905
Automatic burettes, Sofnol
Automatic burettes, Sofnol

10 ml, 0,1 ml, 0,01 ± ml

 
5.060,00 kr
   
1 * 5 Items
 

Borosilicate glass.

  • Rotaflo® PTFE stopcock for grease-free performance and fine drop-wise control
  • Graduations in white enamel
  • Automatic zeroing and easy filling

 : Burettes and bottles must be ordered separately.

 
  • Capacity:
    10 ml
  • Division:
    0,1 ml
  • Tolerance:
    0,01 ± ml
Accessories

 

  •  
    Description
     
     
  • 612-2909
    Spare reservoir bottle, 1000 ml, ØxH: 95x195 mm
       
    1 * 1 Items - 245,00 kr

 

  •  
    Description
     
     
  • 612-2908
    Spare reservoir bottle, 500 ml, ØxH: 75x160 mm
       
    1 * 1 Items - 157,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.