Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium hydroxide 98%, pellets

Sodium hydroxide 98%, pellets

Glass bottle for solids, 500 g
Catalog # A16037.36
Corrosive
Sodium hydroxide 98%, pellets
Catalog # A16037.36
CAS Number:  1310-73-2
Sodium hydroxide 98%, pellets
Sodium hydroxide 98%, pellets

Glass bottle for solids, 500 g

CAS Number:  1310-73-2
Price
433,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,8627
  • Boiling Pt:1390 °C
  • Melting Pt:318 °C
  • Density:2.13
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.