Order Entry
Denmark
Orders LinkContactUsLinkComponent
Formamide ≥99%

Formamide ≥99%

Glass bottle for solids, 500 g
Catalog # A11076.36
Health
Formamide ≥99%
Catalog # A11076.36
CAS Number:  75-12-7
Formamide ≥99%
Formamide ≥99%

Glass bottle for solids, 500 g

CAS Number:  75-12-7
Price
526,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 150°(302°F) Refractive Index: 1.4475 Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941

Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:14,4237
  • Boiling Pt:210 °C
  • Melting Pt:2 °C
  • Density:1.134
  • Flash Pt:150 °C (302 °F)
  • Storage Temperature:Ambient
  • EINECS:200-842-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications