Order Entry
Denmark
Orders LinkContactUsLinkComponent
Syringes

Syringes

Syringe, glass, center nozzle, luer slip, grad. 0,5 ml, 5 ml
Catalog # 613-3002
Supplier:   MURRAY S
Syringes
Catalog # 613-3002
Supplier:  MURRAY S
Syringes
Syringes

Syringe, glass, center nozzle, luer slip, grad. 0,5 ml, 5 ml

Price
78,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Glass syringes, without needles.

  • Centre nozzle with Luer slip fitting
Specifications
  • Capacity:
    5 ml
  • Description:
    Syringe, glass, center nozzle, luer slip, grad. 0,5 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.