Order Entry
Denmark
Orders LinkContactUsLinkComponent
Syringes

Syringes

Syringe, glass, center nozzle, luer slip, grad. 0,2 ml, 2 ml
Catalog # 613-3001
Supplier:   MURRAY S
Syringes
Catalog # 613-3001
Supplier:  MURRAY S
Syringes
Syringes

Syringe, glass, center nozzle, luer slip, grad. 0,2 ml, 2 ml

Price
95,20 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Glass syringes, without needles.

  • Centre nozzle with Luer slip fitting
Specifications
  • Capacity:
    2 ml
  • Description:
    Syringe, glass, center nozzle, luer slip, grad. 0,2 ml